Få mere at vide
5-(Carboxyhydroxymethyl)-2’-O-methyluridine Methyl Ester, TRC
Artikelnummer. 30005949
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30005949 25 mg -
30006147 10 mg -
30009520 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005949 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17275025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2450329-71-0
Molekylær formel C13 H18 N2 O9
Molekylvægt (g/mol) 346.29
Synonym Methyl 2-Hydroxy-2-(1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)acetate
IUPAC navn methyl 2-hydroxy-2-[1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxy-tetrahydrofuran-2-yl]-2,4-dioxo-pyrimidin-5-yl]acetate
SMIL CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C(O)C(=O)OC)C(=O)NC2=O
Tekniske data
Kemisk navn eller materiale 5-(Carboxyhydroxymethyl)-2'-O-methyluridine Methyl Ester
Mængde 25 mg
InChI formel InChI=1S/C13H18N2O9/c1-22-9-8(18)6(4-16)24-11(9)15-3-5(7(17)12(20)23-2)10(19)14-13(15)21/h3,6-9,11,16-18H,4H2,1-2H3,(H,14,19,21)/t6-,7?,8-,9-,11-/m1/s1
Formel vægt 346.1012