Få mere at vide
5,8-Diethyl-7-hydroxy-6-dodecanone Oxime, TRC
Artikelnummer. 30376200
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376200 100 mg
30372586 50 mg
30373859 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30376200 Leverandør Toronto Research Chemicals Leverandørnr. TRCD441795100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6873-77-4
Molekylær formel C16H33NO2
Molekylvægt (g/mol) 271.44
Synonym 5,8-Diethyl-7-hydroxy-6-dodecanonoxime, 5,8-Diethyl-7-hydroxydodecan-6-oxime, 5,8-Diethyl-7-hydroxydodecane-6-oxime, LIX 63
IUPAC navn (7E)-5,8-diethyl-7-hydroxyiminododecan-6-ol
SMIL CCCCC(CC)C(O)\C(=N\O)\C(CC)CCCC
Tekniske data
Kemisk navn eller materiale 5,8-Diethyl-7-hydroxy-6-dodecanone Oxime
Mængde 100 mg
InChI formel InChI=1S/C16H33NO2/c1-5-9-11-13(7-3)15(17-19)16(18)14(8-4)12-10-6-2/h13-14,16,18-19H,5-12H2,1-4H3/b17-15+
Formel vægt 271.2511