Få mere at vide
5,6-Dimethylthiouracil, TRC
Artikelnummer. 30009768
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009768 100 mg
30006105 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009768 Leverandør Toronto Research Chemicals Leverandørnr. TRCD480245100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 28456-54-4
Molekylær formel C6 H8 N2 O S
Molekylvægt (g/mol) 156.21
Synonym Uracil, 5,6-dimethyl-2-thio- (6CI,7CI,8CI), 2,3-Dihydro-5,6-dimethyl-2-thioxo-4(1H)-pyrimidinone, 5,6-Dimethyl-2-thiouracil, 5,6-Dimethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, 5,6-Dimethylthiouracil, NSC 60687, 5,6-Dimethyl-2-thiouracil, DMTU
IUPAC navn 5,6-dimethyl-2-sulfanylidene-1H-pyrimidin-4-one
SMIL CC1=C(C)C(=O)NC(=S)N1
Tekniske data
Kemisk navn eller materiale 5,6-Dimethyl-2-thiouracil
Mængde 100 mg
InChI formel InChI=1S/C6H8N2OS/c1-3-4(2)7-6(10)8-5(3)9/h1-2H3,(H2,7,8,9,10)
Formel vægt 156.0357