Få mere at vide
5-(4-Methylpiperazin-1-yl)pyridin-2-amine, TRC
Artikelnummer. 30480487
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480487 500 mg
30473334 5 g
30481986 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480487 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320895500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 571189-49-6
Molekylær formel C10H16N4
Molekylvægt (g/mol) 192.26
Synonym 5-(4-Methylpiperazin-1-yl)pyridin-2-ylamine, 5-(4-Methyl-1-piperazinyl)-2-pyridinamine, 1-Methyl-4-(6-aminopyridin-3-yl)piperazine, 4-(2-Aminopyridin-5-yl)-1-methylpiperazine
IUPAC navn 5-(4-methylpiperazin-1-yl)pyridin-2-amine
SMIL CN1CCN(CC1)c2ccc(N)nc2
Tekniske data
Kemisk navn eller materiale 5-(4-Methylpiperazin-1-yl)pyridin-2-amine
Mængde 500 mg
InChI formel InChI=1S/C10H16N4/c1-13-4-6-14(7-5-13)9-2-3-10(11)12-8-9/h2-3,8H,4-7H2,1H3,(H2,11,12)
Formel vægt 192.1375