Få mere at vide
5-(3-Bromophenyl)isoxazole, Thermo Scientific™
Artikelnummer. 11480050
Skift visning
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11480050 1 g 1g
11470050 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11480050 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H50580.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7064-33-7
Molekylær formel C31H38N4O4S2
Molekylvægt (g/mol) 594.79
MDL nummer MFCD02183531
InChI nøgle ALUPLTWVMIXEQH-DICXZTSXNA-N
Synonym 5-3-bromophenyl isoxazole, 5-3-bromophenyl-1,2-oxazole, isoxazole,5-3-bromophenyl, zlchem 1341, acmc-209ofi
PubChem CID 2735607
IUPAC navn 5-(3-bromophenyl)-1,2-oxazole
SMIL CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCC3=CC=C(OC)C(OC)=C3)C2=O)=C1N1CC(C)CC(C)C1
Tekniske data
Kemisk navn eller materiale 5-(3-Bromophenyl)isoxazole
Mængde 1 g
Formel vægt 224.06