Få mere at vide
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid, 97%, Thermo Scientific™
Artikelnummer. 15443246
Skift visning
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15443246 250 mg 250mg
15433246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15443246 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H57246.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 199528-28-4
Molekylær formel C14H9F3O2
Molekylvægt (g/mol) 266.219
MDL nummer MFCD03424604
InChI nøgle BJKGBAVGLRYCTK-UHFFFAOYSA-N
Synonym 4'-trifluoromethyl-biphenyl-3-carboxylic acid, 4'-trifluoromethyl-1,1'-biphenyl-3-carboxylic acid, 3-4-trifluoromethyl phenyl benzoic acid, 4'-trifluoromethylbiphenyl-3-carboxylic acid, 4'-trifluoromethyl 1,1'-biphenyl-3-carboxylic acid, 4'-trifluoromethyl biphenyl-3-carboxylic acid, 1,1'-biphenyl-3-carboxylicacid, 4'-trifluoromethyl, pubchem9318, amtda080
PubChem CID 2782718
IUPAC navn 3-[4-(trifluoromethyl)phenyl]benzoic acid
SMIL C1=CC(=CC(=C1)C(=O)O)C2=CC=C(C=C2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Mængde 250 mg
Formel vægt 266.22
Procent renhed 97%