Få mere at vide
4-Phenylbutyric Acid-13C6, TRC
Artikelnummer. 30133097
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133097 100 mg
30008358 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133097 Leverandør Toronto Research Chemicals Leverandørnr. TRCP319613100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346602-38-7
Molekylær formel C413C6H12O2
Molekylvægt (g/mol) 170.16
Synonym Benzene-13C6-butanoic Acid, γ-Phenyl-13C6-butanoic Acid;
SMIL [H][13C]1=[13C](CCCC(O)=O)[13C]([H])=[13C]([H])[13C]([H])=[13C]1[H]
Tekniske data
Kemisk navn eller materiale 4-Phenylbutyric Acid-13C6
Mængde 100 mg
InChI formel InChI=1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)/i1+1,2+1,3+1,5+1,6+1,9+1
Formel vægt 170.1039