Få mere at vide
4-Oxo-1,4-dihydroquinoline Carboxylic Acid, TRC
Artikelnummer. 30422908
Skift visning
Mængde:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422908 50 g
30418753 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30422908 Leverandør Toronto Research Chemicals Leverandørnr. TRCO85540050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13721-01-2
Molekylær formel C10H7NO3
Molekylvægt (g/mol) 189.17
Synonym 1,4-Dihydro-4-oxo-3-quinolinecarboxylic Acid, 4-Quinolone-3-carboxylic Acid
IUPAC navn 4-oxo-1H-quinoline-3-carboxylic acid
SMIL OC(=O)C1=CNc2ccccc2C1=O
Tekniske data
Kemisk navn eller materiale 4-Oxo-1,4-dihydroquinoline Carboxylic Acid
Mængde 50 g
InChI formel InChI=1S/C10H7NO3/c12-9-6-3-1-2-4-8(6)11-5-7(9)10(13)14/h1-5H,(H,11,12)(H,13,14)
Formel vægt 189.0426