Få mere at vide
4-Methylbenzotriazole, TRC
Artikelnummer. 30133661
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133661 1 g
30137657 500 mg
17972505 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133661 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2887801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29878-31-7
Molekylær formel C7 H7 N3
Molekylvægt (g/mol) 133.15
Synonym 4-Methylbenzotriazole, 1H-Benzotriazole, 7-methyl-, 1H-Benzotriazole, 4-methyl- (8CI,9CI), Benzotriazole, 4-methyl- (6CI), 7-Methyl-1H-benzotriazole, 4-Methyl-1,2,3-benzotriazole, 4-Methyl-1H-benzo[d][1,2,3]triazole, 4-Methyl-1H-benzotriazole, 4-Methylbenzotriazole, 1H-Benzotriazole, 7-methyl-, 1H-Benzotriazole, 4-methyl- (8CI,9CI), Benzotriazole, 4-methyl- (6CI), 7-Methyl-1H-benzotriazole, 4-Methyl-1,2,3-benzotriazole, 4-Methyl-1H-benzo[d][1,2,3]triazole, 4-Methyl-1H-benzotriazole
IUPAC navn 4-methyl-1H-benzotriazole
SMIL Cc1cccc2[nH]nnc12
Tekniske data
Kemisk navn eller materiale 4-Methylbenzotriazole
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C7H7N3/c1-5-3-2-4-6-7(5)9-10-8-6/h2-4H,1H3,(H,8,9,10)
Formel vægt 133.064
Procent renhed >95%