Få mere at vide
4-(Methylamino)-1-(3-pyridyl)-1-butanone Dihydrochloride, TRC
Artikelnummer. 30007042
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007042 1 g
17912745 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007042 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2870001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66093-90-1
Molekylær formel C10 H14 N2 O . 2 Cl H
Molekylvægt (g/mol) 251.15
Synonym Pseudooxynicotine Dihydrochloride, 1-Butanone, 4-(methylamino)-1-(3-pyridinyl)-, hydrochloride (1:2), 1-Butanone, 4-(methylamino)-1-(3-pyridinyl)-, dihydrochloride (9CI), 1-Butanone, 4-methylamino-1-(3-pyridyl)-, dihydrochloride
IUPAC navn 4-(methylamino)-1-pyridin-3-ylbutan-1-one;dihydrochloride
SMIL Cl.Cl.CNCCCC(=O)c1cccnc1
Tekniske data
Kemisk navn eller materiale 4-(Methylamino)-1-(3-pyridyl)-1-butanone Dihydrochloride
Mængde 1 g
InChI formel InChI=1S/C10H14N2O.2ClH/c1-11-6-3-5-10(13)9-4-2-7-12-8-9;;/h2,4,7-8,11H,3,5-6H2,1H3;2*1H
Formel vægt 250.064