Få mere at vide
4-Methyl-1-nitrosopiperidine, TRC
Artikelnummer. 30481586
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481586 500 mg
30471540 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30481586 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325794500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15104-03-7
Molekylær formel C6H12N2O
Molekylvægt (g/mol) 128.17
Synonym 1-Nitroso-4-pipecoline, 4-Methyl-N-nitrosopiperidine
IUPAC navn 4-methyl-1-nitrosopiperidine
SMIL CC1CCN(CC1)N=O
Tekniske data
Kemisk navn eller materiale 4-Methyl-1-nitrosopiperidine
Mængde 500 mg
InChI formel InChI=1S/C6H12N2O/c1-6-2-4-8(7-9)5-3-6/h6H,2-5H2,1H3
Formel vægt 128.095