missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Methyl-1,3-phenylenediamine, Sigma-Aldrich™

Artikelnummer. 15868980
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15868980 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15868980 Leverandør Sigma Aldrich Leverandørnr. 8032690250
 Begrænset vare
Forventet forsendelse: 24-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-80-7
Grad Research
Molekylær formel 4-(CH3)C6H3-1,3-(NH2)2
Molekylvægt (g/mol) 122.17

Specifications

Smeltepunkt 99°C
Tæthed 1.26 g/cm3 at 20°C
MDL nummer MFCD00007804
Mængde 250 g
Synonym 4-Methyl-1,3-phenylenediamine, 2,4-Diaminotoluene, 2,4-Toluylenediamine, 4-Methyl-1,3-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.