Få mere at vide
4'-Methoxy PhIP, TRC
Artikelnummer. 30483724
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483724 10 mg
30480048 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483724 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26640010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797911-00-2
Molekylær formel C14H14N4O
Molekylvægt (g/mol) 254.29
Synonym 4-(2-Amino-1-methyl-1H-imidazo[4,5-b]pyridin-6-yl)phenol Methyl Ether, 2-Amino-4'-methoxy-1-methyl-6-phenylimidazo[4,5-b]pyridine
SMIL COc1ccc(cc1)c2cnc3nc(N)n(C)c3c2
Tekniske data
Kemisk navn eller materiale 4'-Methoxy PhIP
Mængde 10 mg
InChI formel InChI=1S/C14H14N4O/c1-18-12-7-10(8-16-13(12)17-14(18)15)9-3-5-11(19-2)6-4-9/h3-8H,1-2H3,(H2,15,16,17)