Få mere at vide
4-Methoxy Clonidine, TRC
Artikelnummer. 30467595
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30467595 25 mg -
30483707 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30467595 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26214025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 65936-24-5
Molekylær formel C10H11Cl2N3O
Molekylvægt (g/mol) 260.12
Synonym N-(2,6-dichloro-4-methoxyphenyl)-4,5-dihydro-1H-imidazol-2-amine
IUPAC navn N-(2,6-dichloro-4-methoxyphenyl)-4,5-dihydro-1H-imidazol-2-amine
SMIL COc1cc(Cl)c(NC2=NCCN2)c(Cl)c1
Tekniske data
Kemisk navn eller materiale 4-Methoxy Clonidine
Mængde 25 mg
InChI formel InChI=1S/C10H11Cl2N3O/c1-16-6-4-7(11)9(8(12)5-6)15-10-13-2-3-14-10/h4-5H,2-3H2,1H3,(H2,13,14,15)
Formel vægt 259.0279