Få mere at vide
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanol Hydrochloride, TRC
Artikelnummer. 30133284
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133284 500 mg
30003659 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133284 Leverandør Toronto Research Chemicals Leverandørnr. TRCM262335500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 96300-88-8
Molekylær formel C9 H13 N O2 . Cl H
Molekylvægt (g/mol) 203.67
Synonym 2-Pyridinemethanol, 4-methoxy-3,5-dimethyl-, hydrochloride (1:1), 2-Pyridinemethanol, 4-methoxy-3,5-dimethyl-, hydrochloride (9CI), 2-Hydroxymethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride
IUPAC navn (4-methoxy-3,5-dimethylpyridin-2-yl)methanol;hydrochloride
SMIL Cl.COc1c(C)cnc(CO)c1C
Tekniske data
Kemisk navn eller materiale (4-Methoxy-3,5-dimethylpyridin-2-yl)methanol hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C9H13NO2.ClH/c1-6-4-10-8(5-11)7(2)9(6)12-3;/h4,11H,5H2,1-3H3;1H
Formel vægt 203.0713