missing translation for 'onlineSavingsMsg'
Få mere at vide

4′-Hydroxypropiophenone, Sigma-Aldrich™

Artikelnummer. 15811121
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15811121 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Artikelnummer. 15811121 Leverandør Sigma Aldrich Leverandørnr. 8206920100
 Begrænset vare
Forventet forsendelse: 26-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 70-70-2
Smeltepunkt 148°C to 151°C
Tæthed 1.09 g/cm3 at 20°C
Molekylær formel 4-(OH)C6H4COC2H5
MDL nummer MFCD00002361
Mængde 100 g
Synonym 4′-Hydroxypropiophenone, 4′-Hydroxypropiophenone
Molekylvægt (g/mol) 150.17
Grad Research
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.