Få mere at vide
(4-Chloro-3-methylphenyl)hydrazine Hydrochloride, TRC
Artikelnummer. 30367605
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367605 250 mg
30352615 100 mg
30349146 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367605 Leverandør Toronto Research Chemicals Leverandørnr. TRCC371320250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 221687-08-7
Molekylær formel C7H9ClN2.HCl
Molekylvægt (g/mol) 193.07
Synonym (4-Chloro-3-methylphenyl)hydrazine Monohydrochloride
IUPAC navn (4-chloro-3-methylphenyl)hydrazine;hydrochloride
SMIL Cl.Cc1cc(NN)ccc1Cl
Tekniske data
Kemisk navn eller materiale (4-Chloro-3-methylphenyl)hydrazine Hydrochloride
Mængde 250 mg
InChI formel InChI=1S/C7H9ClN2.ClH/c1-5-4-6(10-9)2-3-7(5)8;/h2-4,10H,9H2,1H3;1H
Formel vægt 192.0221