missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™

Artikelnummer. 11485900
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11485900 1 g 1g
11495900 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11485900 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H26512.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-2-fluorophenylacetic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 194240-75-0
Molekylær formel C8H6ClFO2
Molekylvægt (g/mol) 188.58
MDL nummer MFCD03095402
InChI nøgle WKAZXGAJEGPUAY-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorophenylacetic acid, 2-4-chloro-2-fluorophenyl acetic acid, 4-chloro-2-fluorophenyl acetic acid, 4-chloro-2-fluorophenylaceticacid, rarechem al bo 0928, pubchem4292, acmc-20a3w8, ksc541i7t, 4-chlor-2-flour phenyactic acid, 4-chloro-2-fluoro-phenyl acetic acid
PubChem CID 2757563
IUPAC navn 2-(4-chloro-2-fluorophenyl)acetic acid
SMIL OC(=O)CC1=CC=C(Cl)C=C1F

Tekniske data

Kemisk navn eller materiale 4-Chloro-2-fluorophenylacetic acid
Mængde 1 g
Opløselighedsinformation Slightly soluble in water.
Formel vægt 188.59
Procent renhed 97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.