Få mere at vide
4-Chloro-2-fluorobenzyl bromide, 97%, Thermo Scientific™
Artikelnummer. 11336939
Skift visning
Mængde:
1 g
10 g
Pakningsstørrelse:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11336939 10 g 10g
11326939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11336939 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H26814.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71916-82-0
Molekylær formel C7H5BrClF
Molekylvægt (g/mol) 223.469
MDL nummer MFCD00831105
InChI nøgle UFCSSWZQROEFBZ-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-chloro-2-fluorobenzene, 4-chloro-2-fluorobenzyl bromide, 2-fluoro-4-chlorobenzyl bromide, 4-chloro-2-fluorobenzylbromide, alpha-bromo-4-chloro-2-fluorotoluene, benzene, 1-bromomethyl-4-chloro-2-fluoro, pubchem4897, 1-bromomethyl-4-chloro-2-fluoro-benzene, acmc-209okn
PubChem CID 2736551
IUPAC navn 1-(bromomethyl)-4-chloro-2-fluorobenzene
SMIL C1=CC(=C(C=C1Cl)F)CBr
Tekniske data
Kemisk navn eller materiale 4-Chloro-2-fluorobenzyl bromide
Kogepunkt 96°C to 98°C (15 mmHg)
Brydningsindeks 1.568
Mængde 10 g
FN nummer UN3265
Formel vægt 223.48
Procent renhed 97%