Få mere at vide
(4-Carboxy-3-ethoxy)phenyl Acetic Acid, TRC
Artikelnummer. 30353871
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353871 25 mg
30367389 5 mg
30365996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30353871 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17796025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 220438-80-2
Molekylær formel C11 H12 O5
Molekylvægt (g/mol) 224.21
Synonym Benzeneacetic acid, 4-carboxy-3-ethoxy-, 4-Carboxy-3-ethoxybenzeneacetic acid, 4-Carboxymethyl-2-ethoxybenzoic acid, Repaglinide Imp. A (EP)
IUPAC navn 4-(carboxymethyl)-2-ethoxybenzoic acid
SMIL CCOc1cc(CC(=O)O)ccc1C(=O)O
Tekniske data
Kemisk navn eller materiale (4-Carboxy-3-ethoxy)phenyl Acetic Acid
Mængde 25 mg
InChI formel InChI=1S/C11H12O5/c1-2-16-9-5-7(6-10(12)13)3-4-8(9)11(14)15/h3-5H,2,6H2,1H3,(H,12,13)(H,14,15)
Formel vægt 224.0685