Få mere at vide
4-(Butylamino)benzoic Acid, TRC
Artikelnummer. 30133753
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133753 10 g
30138368 5 g
17913935 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133753 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68944510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4740-24-3
Molekylær formel C11 H15 N O2
Molekylvægt (g/mol) 193.24
Synonym 4-(Butylamino)benzoic acid, Tetracaine Hydrochloride Imp. B (EP), Benzoic acid, 4-(butylamino)-, Benzoic acid, p-(butylamino)- (6CI,7CI,8CI), NSC 44300, p-Butylaminobenzoic acid
IUPAC navn 4-(butylamino)benzoic acid
SMIL CCCCNc1ccc(cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale 4-(Butylamino)benzoic Acid
Mængde 10 g
InChI formel InChI=1S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12H,2-3,8H2,1H3,(H,13,14)
Formel vægt 193.1103