Få mere at vide
4-Amino-5-chloro-2,3-dihydro-7-benzofurancarboxylic Acid, TRC
Artikelnummer. 30137880
Skift visning
Mængde:
2.5 g
5 g
25 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137880 5 g -
30138534 25 g -
30007974 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137880 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6039705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123654-26-2
Molekylær formel C9H8ClNO3
Molekylvægt (g/mol) 213.62
Synonym 4-Amino-5-chloro-2,3-dihydrobenzo[b]furan-7-carboxylic Acid, Prucalopride Impurity
IUPAC navn 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
SMIL Nc1c(Cl)cc(C(=O)O)c2OCCc12
Tekniske data
Kemisk navn eller materiale 4-Amino-5-chloro-2,3-dihydro-7-benzofurancarboxylic Acid
Mængde 5 g
InChI formel InChI=1S/C9H8ClNO3/c10-6-3-5(9(12)13)8-4(7(6)11)1-2-14-8/h3H,1-2,11H2,(H,12,13)
Formel vægt 213.0193