Få mere at vide
4,6-Dimethyl-2-(methylsulfonyl)pyrimidine, TRC
Artikelnummer. 30134849
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134849 10 g
17991745 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134849 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44693010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35144-22-0
Molekylær formel C7H10N2O2S
Molekylvægt (g/mol) 186.23
Synonym 2-(Methylsulfonyl)-4,6-dimethylpyrimidine, 2-Methanesulfonyl-4,6-dimethylpyrimidine
SMIL Cc1cc(C)nc(n1)S(=O)(=O)C
Tekniske data
Kemisk navn eller materiale 4,6-Dimethyl-2-(methylsulfonyl)pyrimidine
Mængde 10 g
InChI formel InChI=1S/C7H10N2O2S/c1-5-4-6(2)9-7(8-5)12(3,10)11/h4H,1-3H3
Formel vægt 186.0463