Få mere at vide
4,5-Dichloroveratrole, TRC
Artikelnummer. 30372182
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372182 100 mg
30379332 50 mg
30388521 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30372182 Leverandør Toronto Research Chemicals Leverandørnr. TRCD437580100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2772-46-5
Molekylær formel C8H8O2Cl2
Molekylvægt (g/mol) 207.05
Synonym 4,5-Dichloroveratrole, Benzene, 1,2-dichloro-4,5-dimethoxy-, Veratrole, 4,5-dichloro- (6CI), 1,2-Dichloro-4,5-dimethoxybenzene, 4,5-Dichloro-1,2-dimethoxybenzene, 4,5-Dichloroveratrole
IUPAC navn 1,2-dichloro-4,5-dimethoxybenzene
SMIL COc1cc(Cl)c(Cl)cc1OC
Tekniske data
Kemisk navn eller materiale 4,5-Dichloroveratrole
Mængde 100 mg
InChI formel InChI=1S/C8H8Cl2O2/c1-11-7-3-5(9)6(10)4-8(7)12-2/h3-4H,1-2H3
Formel vægt 205.9901