Få mere at vide
4,4'-Dichlorobenzhydrol, 98%, Thermo Scientific™
Beskrivelse
Denne Thermo Scientific mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific .
Applications
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4,4'-Dichlorobenzhydrol |
| Smeltepunkt | 91°C to 93°C |
| Mængde | 100 g |
| Beilstein | 1878717 |
| Opløselighedsinformation | Insoluble in water. |
| Formel vægt | 253.13 |
| Procent renhed | 98% |