Få mere at vide
4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, TRC
Artikelnummer. 30364358
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364358 1 g
30367356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364358 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3778601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36159-73-6
Molekylær formel C16H13ClO
Molekylvægt (g/mol) 256.73
Synonym 1(2H)-Naphthalenone, 4-(4-chlorophenyl)-3,4-dihydro-, 4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
IUPAC navn 4-(4-chlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
SMIL Clc1ccc(cc1)C2CCC(=O)c3ccccc23
Tekniske data
Kemisk navn eller materiale 4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
Mængde 1 g
InChI formel InChI=1S/C16H13ClO/c17-12-7-5-11(6-8-12)13-9-10-16(18)15-4-2-1-3-14(13)15/h1-8,13H,9-10H2
Formel vægt 256.0655