Få mere at vide
4-[4-(4-Chlorophenyl)-4-hydroxypiperidine]-4-defluorohaloperidol Decanoate, TRC
Artikelnummer. 30004086
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004086 50 mg
30008200 5 mg
30009874 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004086 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37857050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797008-46-8
Molekylær formel C42 H54 Cl2 N2 O4
Molekylvægt (g/mol) 721.8
Synonym 4-(4-Chlorophenyl)-1-(4-(4-(4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl)phenyl)-4-oxobutyl)piperidin-4-yl decanoate, Haloperidol Decanoate EP Impurity D;
IUPAC navn [4-(4-chlorophenyl)-1-[4-[4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidyl]phenyl]-4-oxo-butyl]-4-piperidyl] decanoate
SMIL CCCCCCCCCC(=O)OC1(CCN(CCCC(=O)c2ccc(cc2)N3CCC(O)(CC3)c4ccc(Cl)cc4)CC1)c5ccc(Cl)cc5
Tekniske data
Kemisk navn eller materiale 4-[4-(4-Chlorophenyl)-4-hydroxypiperidine]-4-defluorohaloperidol Decanoate
Mængde 50 mg
InChI formel InChI=1S/C42H54Cl2N2O4/c1-2-3-4-5-6-7-8-11-40(48)50-42(35-16-20-37(44)21-17-35)26-29-45(30-27-42)28-9-10-39(47)33-12-22-38(23-13-33)46-31-24-41(49,25-32-46)34-14-18-36(43)19-15-34/h12-23,49H,2-11,24-32H2,1H3
Formel vægt 720.3461