Få mere at vide
4-[2-(Diethylamino)ethoxy]benzophenone, TRC
Artikelnummer. 30134379
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134379 500 mg
30003971 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134379 Leverandør Toronto Research Chemicals Leverandørnr. TRCD470920500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 796-77-0
Molekylær formel C19 H23 N O2
Molekylvægt (g/mol) 297.39
Synonym [4-[2-(Diethylamino)ethoxy]phenyl]phenylmethanone, Benzophenone, 4-[2-(diethylamino)ethoxy]- (6CI,7CI,8CI), 4-(β-Diethylaminoethoxy)benzophenone, 4-[2-(Diethylamino)ethoxy]benzophenone
IUPAC navn [4-[2-(diethylamino)ethoxy]phenyl]-phenylmethanone
SMIL CCN(CC)CCOc1ccc(cc1)C(=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale (4-[2-(Diethylamino)ethoxy]phenyl)phenylmethanone
Mængde 500 mg
InChI formel InChI=1S/C19H23NO2/c1-3-20(4-2)14-15-22-18-12-10-17(11-13-18)19(21)16-8-6-5-7-9-16/h5-13H,3-4,14-15H2,1-2H3
Formel vægt 297.1729