Få mere at vide
[[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]methyl]oxirane, TRC
Artikelnummer. 30133288
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133288 250 mg
30012372 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133288 Leverandør Toronto Research Chemicals Leverandørnr. TRCM304765250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66722-57-4
Molekylær formel C15 H22 O4
Molekylvægt (g/mol) 266.33
Synonym 2-[4-(2-Isopropoxyethoxymethyl)phenoxymethyl]oxirane
IUPAC navn 2-[[4-(2-propan-2-yloxyethoxymethyl)phenoxy]methyl]oxirane
SMIL CC(C)OCCOCc1ccc(OCC2CO2)cc1
Tekniske data
Kemisk navn eller materiale 2-[4-(2-Isopropoxyethoxymethyl)phenoxymethyl]oxirane
Mængde 250 mg
InChI formel InChI=1S/C15H22O4/c1-12(2)17-8-7-16-9-13-3-5-14(6-4-13)18-10-15-11-19-15/h3-6,12,15H,7-11H2,1-2H3
Formel vægt 266.1518