Få mere at vide
3Alpha,12Beta-Dihydroxycholanoic Acid, TRC
Artikelnummer. 30134126
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134126 100 mg
30009322 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134126 Leverandør Toronto Research Chemicals Leverandørnr. TRCD447440100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 570-62-7
Molekylær formel C24H40O4
Molekylvægt (g/mol) 392.57
Synonym 3α,12β-Dihydroxy-5β-cholan-24-oic Acid, 3α,12β-Dihydroxy-5β-cholanic Acid, 12-Epideoxycholic Acid, 12-epi-Deoxycholic Acid, 3α,12β-Dihydroxy-5β-cholan-24-oic Acid, 3α,12β-Dihydroxy-5β-cholanic Acid, 3α,12β-Dihydroxy-5β-cholanoic Acid, (R)-4-((3R,5R,8R,9S,10S,12R,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic Acid
SMIL OC(CC[C@@H](C)[C@@]1([H])CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@@H](O)[C@@]21C)=O
Tekniske data
Kemisk navn eller materiale 3alpha,12beta-Dihydroxycholanoic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21-,23+,24-/m1/s1
Formel vægt 392.2927
Procent renhed >95%