Få mere at vide
3-Nitro-4-acetamidophenol, TRC
Artikelnummer. 30476662
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476662 5 g
30472549 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30476662 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4902755G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7403-75-0
Molekylær formel C8 H8 N2 O4
Molekylvægt (g/mol) 196.16
IUPAC navn N-(4-hydroxy-2-nitrophenyl)acetamide
SMIL CC(=O)Nc1ccc(O)cc1[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 3-Nitro-4-acetamidophenol
Mængde 5 g
InChI formel InChI=1S/C8H8N2O4/c1-5(11)9-7-3-2-6(12)4-8(7)10(13)14/h2-4,12H,1H3,(H,9,11)
Formel vægt 196.0484