missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Isopropoxybenzoic Acid, TRC

Artikelnummer. 30407695
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407695 100 mg
30407543 500 mg
30405135 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30407695 Leverandør Toronto Research Chemicals Leverandørnr. TRCI918195100MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60772-67-0
Molekylær formel C10H12O3
Molekylvægt (g/mol) 180.2
Synonym 3-(1-Methylethoxy)benzoic Acid, 3-Isopropoxybenzoic Acid
IUPAC navn 3-propan-2-yloxybenzoic acid
SMIL CC(C)Oc1cccc(c1)C(=O)O

Specifications

Kemisk navn eller materiale 3-Isopropoxybenzoic Acid
Mængde 100 mg
InChI formel InChI=1S/C10H12O3/c1-7(2)13-9-5-3-4-8(6-9)10(11)12/h3-7H,1-2H3,(H,11,12)
Formel vægt 180.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.