Få mere at vide
3-hydroxycyclohexan-1-one, TRC
Artikelnummer. 30138753
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138753 50 mg
30133981 10 mg
30012679 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138753 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95494050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 823-19-8
Molekylær formel C6H10O2
Molekylvægt (g/mol) 114.14
IUPAC navn 3-hydroxycyclohexan-1-one
SMIL OC1CCCC(=O)C1
Tekniske data
Kemisk navn eller materiale 3-hydroxycyclohexan-1-one
Mængde 50 mg
InChI formel InChI=1S/C6H10O2/c7-5-2-1-3-6(8)4-5/h5,7H,1-4H2
Formel vægt 114.0681