Få mere at vide
3-Hydroxy Detomidine Hydrochloride, TRC
Artikelnummer. 30006790
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006790 1 mg
30010464 5 mg
30008556 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006790 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8253401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794759-72-0
Molekylær formel C12 H14 N2 O . Cl H
Molekylvægt (g/mol) 238.71
Synonym 3-(1H-Imidazol-5-ylmethyl)-2-methylbenzenemethanol Hydrochloride, 3-(1H-Imidazol-4-ylmethyl)-2-methylbenzenemethanol Hydrochloride;
IUPAC navn [3-(1H-imidazol-4-ylmethyl)-2-methylphenyl]methanol;hydrochloride
SMIL Cl.Cc1c(CO)cccc1Cc2c[nH]cn2
Tekniske data
Kemisk navn eller materiale 3-Hydroxy Detomidine Hydrochloride
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C12H14N2O.ClH/c1-9-10(3-2-4-11(9)7-15)5-12-6-13-8-14-12;/h2-4,6,8,15H,5,7H2,1H3,(H,13,14);1H
Formel vægt 238.0873
Procent renhed >95%