missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™

Catalog Number 15473447
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886499-13-4
C8H7F4NO
209.14
MFCD06660207
LKKOOHKPOMOOAI-UHFFFAOYSA-N
3-fluoro-4-trifluoromethoxy benzylamine, 3-fluoro-4-trifluoromethoxy-benzylamine, 3-fluoro-4-trifluoromethoxy phenyl methanamine, benzenemethanamine,3-fluoro-4-trifluoromethoxy, 1-3-fluoro-4-trifluoromethoxy phenyl methanamine, 3-fluoro-4-trifluoromethoxybenzylamine, 3-fluoro-4-trifluoromethoxy phenyl methylamine
17750674
[3-fluoro-4-(trifluoromethoxy)phenyl]methanamine
NCC1=CC(F)=C(OC(F)(F)F)C=C1
This item is not returnable. View return policy
886499-13-4
C8H7F4NO
209.14
MFCD06660207
LKKOOHKPOMOOAI-UHFFFAOYSA-N
3-fluoro-4-trifluoromethoxy benzylamine, 3-fluoro-4-trifluoromethoxy-benzylamine, 3-fluoro-4-trifluoromethoxy phenyl methanamine, benzenemethanamine,3-fluoro-4-trifluoromethoxy, 1-3-fluoro-4-trifluoromethoxy phenyl methanamine, 3-fluoro-4-trifluoromethoxybenzylamine, 3-fluoro-4-trifluoromethoxy phenyl methylamine
17750674
[3-fluoro-4-(trifluoromethoxy)phenyl]methanamine
NCC1=CC(F)=C(OC(F)(F)F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 209.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'