Få mere at vide
3-Fluoro-4-iodotoluene, 97%, Thermo Scientific™
Artikelnummer. 15480368
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15480368 1 g 1g
15490368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15480368 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H53491.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 452-79-9
Molekylær formel C7H6FI
Molekylvægt (g/mol) 236.028
MDL nummer MFCD00143162
InChI nøgle XVHYSMVSLDIUCM-UHFFFAOYSA-N
Synonym 3-fluoro-4-iodotoluene, 2-fluoro-1-iodo-4-methyl-benzene, benzene, 2-fluoro-1-iodo-4-methyl, pubchem1621, ksc493m8l, 4-methyl-2-fluoro-1-iodobenzene, buttpark 148\01-16
PubChem CID 2774524
IUPAC navn 2-fluoro-1-iodo-4-methylbenzene
SMIL CC1=CC(=C(C=C1)I)F
Tekniske data
Kemisk navn eller materiale 3-Fluoro-4-iodotoluene
Tæthed 1.803
Kogepunkt 138°C to 139°C (9 mmHg)
Flammepunkt 92°C (198°F)
Brydningsindeks 1.581
Mængde 1 g
Følsomhed Light sensitive
Formel vægt 236.03
Procent renhed 97%