missing translation for 'onlineSavingsMsg'
Learn More
3-Ethyl-4-methyl-3-pyrrolin-2-one, 98% Product Code.: 15441198

3-Ethyl-4-methyl-3-pyrrolin-2-one, 98%

Product Code. 15441198
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15441198

Brand: Thermo Scientific Alfa Aesar H55065.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 766-36-9 | C7H11NO | 125.17 g/mol

3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-36-9
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.17
MDL Number MFCD00173861
InChI Key YCTNTSVMJWIYTQ-UHFFFAOYSA-N
Synonym 3-ethyl-4-methyl-3-pyrrolin-2-one, 3-ethyl-4-methyl-3-pyrroline-2-one, 3-ethyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 3-ethyl-4-methyl-1h-pyrrol-2 5h-one, 1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one, 2h-pyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl, 3-ethyl-4-methyl-2-oxo-3-pyrroline, 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl-2h-pyrrol-2-one, pubchem12392Show MoreShow Less
PubChem CID 854146
SMILES CCC1=C(C)CNC1=O

Specifications

Melting Point 100°C to 104°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
IUPAC Name 4-ethyl-3-methyl-1,2-dihydropyrrol-5-one
Formula Weight 125.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-4-methyl-3-pyrrolin-2-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberBF220501 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55065.03,H55065.MD,H55065.06
Lot NumberR18E034 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55065.03,H55065.MD,H55065.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Ethyl-4-methyl-3-pyrrolin-2-one, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.