missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Deoxy-1,2:5,6-di-O-isopropylidene-Alpha-D-glucofuranose, TRC

Artikelnummer. 30179434
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30179434 2.5 g 2.5g
30179472 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30179434 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2328152.5G

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4613-62-1
Molekylær formel C12H20O5
Molekylvægt (g/mol) 244.29
Synonym 3-Deoxy-1,2:5,6-di-O-isopropyliden-alpha-D-ribo Hexafuranose;
SMIL CC1(C)OC[C@@H](O1)[C@@H]2C[C@H]3OC(C)(C)O[C@H]3O2

Tekniske data

Kemisk navn eller materiale 3-Deoxy-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose
Mængde 2.5 g
InChI formel InChI=1S/C12H20O5/c1-11(2)13-6-9(16-11)7-5-8-10(14-7)17-12(3,4)15-8/h7-10H,5-6H2,1-4H3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.