Få mere at vide
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid, 98%, Thermo Scientific™
Artikelnummer. 11488576
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11488576 5 g 5g
11478576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11488576 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A15988.06
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 175135-56-5
Molekylær formel C9H7ClNO6
Molekylvægt (g/mol) 260.61
MDL nummer MFCD00052955
InChI nøgle DQYMZXRGXUJHLS-UHFFFAOYSA-M
Synonym 3-chloro-2,6-dimethoxy-5-nitrobenzoic acid, benzoic acid,3-chloro-2,6-dimethoxy-5-nitro, maybridge1_001816, acmc-20an7i, 5-chloro-2,6-dimethoxy-3-nitrobenzoic acid
PubChem CID 2774753
IUPAC navn 3-chloro-2,6-dimethoxy-5-nitrobenzoate
SMIL COC1=C(Cl)C=C(C(OC)=C1C([O-])=O)[N+]([O-])=O
Tekniske data
Kemisk navn eller materiale 3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
Smeltepunkt 129°C to 132°C
Mængde 5 g
Formel vægt 261.62
Procent renhed 98%