Få mere at vide
3-Bromo-5-methylbenzeneboronic acid, 97%, Thermo Scientific™
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Bromo-5-methylbenzeneboronic acid |
| Smeltepunkt | 276°C to 278°C |
| Mængde | 5 g |
| Formel vægt | 214.85 |
| Procent renhed | 97% |