Få mere at vide
3-Beta-Ursodeoxycholic Acid, TRC
Artikelnummer. 30006614
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006614 50 mg
17903965 10 mg
17973125 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006614 Leverandør Toronto Research Chemicals Leverandørnr. TRCU85000550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78919-26-3
Molekylær formel C24 H40 O4
Molekylvægt (g/mol) 392.57
Synonym 3β,7β-Dihydroxy-5β-cholan-24-oic Acid, (3β,5β,7β)-3,7-dihydroxy-cholan-24-oic acid, 3β,7β-Dihydroxy-5β-cholan-24-oic acid, 3β,7β-Dihydroxy-5β-cholanic acid, 3β,7β-Dihydroxy-5β-cholanoic acid, 3β-Ursodeoxycholic acid, Isoursodeoxycholic acid
IUPAC navn (4R)-4-[(3S,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 3beta-Ursodeoxycholic Acid
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16+,17-,18+,19+,20+,22+,23+,24-/m1/s1
Formel vægt 392.2927
Procent renhed >95%