Få mere at vide
3-Benzylidenecamphor, TRC
Artikelnummer. 30009066
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009066 250 mg
30007018 1 g
17914675 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009066 Leverandør Toronto Research Chemicals Leverandørnr. TRCB280048250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15087-24-8
Molekylær formel C17 H20 O
Molekylvægt (g/mol) 240.34
Synonym 2-Bornanone, 3-benzylidene- (8CI), 1,7,7-Trimethyl-3-(phenylmethylene)bicyclo[2.2.1]heptan-2-one, 3-Benzylidenebornan-2-one, 3-Benzylidenecamphor, Benzylidenecamphor, Eusolex 6900, Ultracyd, Ultren BK, Unisol S 22
IUPAC navn 2-benzylidene-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMIL CC1(C)C2CCC1(C)C(=O)C2=Cc3ccccc3
Tekniske data
Kemisk navn eller materiale 3-Benzylidenecamphor
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C17H20O/c1-16(2)14-9-10-17(16,3)15(18)13(14)11-12-7-5-4-6-8-12/h4-8,11,14H,9-10H2,1-3H3
Formel vægt 240.1514
Procent renhed >95%