Få mere at vide
3,5-Disulfobenzoic Acid, TRC
Artikelnummer. 30354304
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354304 500 mg
30366717 1 g
30349642 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30354304 Leverandør Toronto Research Chemicals Leverandørnr. TRCD121485500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 121-48-2
Molekylær formel C7H6O8S2
Molekylvægt (g/mol) 282.24
IUPAC navn 3,5-disulfobenzoic acid
SMIL OC(=O)c1cc(cc(c1)S(=O)(=O)O)S(=O)(=O)O
Tekniske data
Kemisk navn eller materiale 3,5-Disulfobenzoic Acid
Mængde 500 mg
InChI formel InChI=1S/C7H6O8S2/c8-7(9)4-1-5(16(10,11)12)3-6(2-4)17(13,14)15/h1-3H,(H,8,9)(H,10,11,12)(H,13,14,15)
Formel vægt 281.9504