Få mere at vide
3,5-Dimethyl-1-adamantanol, TRC
Artikelnummer. 30134818
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134818 250 mg
30005241 100 mg
30003595 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134818 Leverandør Toronto Research Chemicals Leverandørnr. TRCD470580250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 707-37-9
Molekylær formel C12 H20 O
Molekylvægt (g/mol) 180.29
Synonym 1-Hydroxy-3,5-dimethyladamantane, Memantine Related Compound B (USP)
IUPAC navn 3,5-dimethyladamantan-1-ol
SMIL CC12CC3CC(C)(C1)CC(O)(C3)C2
Tekniske data
Kemisk navn eller materiale 1-Hydroxy-3,5-dimethyladamantane
Renhedsgrad noter GC
Mængde 250 mg
InChI formel InChI=1S/C12H20O/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9,13H,3-8H2,1-2H3
Formel vægt 180.1514
Procent renhed >95%