missing translation for 'onlineSavingsMsg'
Få mere at vide

3',5'-Dibenzyloxyacetophenone, 98%, Thermo Scientific™

Artikelnummer. 11494157
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
50 g
250 g
Pakningsstørrelse:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11494157 10 g 10g
11404167 50 g 50g
11414167 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11494157 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A19614.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dibenzyloxyacetophenone is used in the synthesis of enantiomerically pure B-ring modified analogs of (-)-epicatechin gallate.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 28924-21-2
Molekylær formel C22H20O3
Molekylvægt (g/mol) 332.399
MDL nummer MFCD00004777
InChI nøgle KOJXGMJOTRYLBD-UHFFFAOYSA-N
PubChem CID 120098
IUPAC navn 1-[3,5-bis(phenylmethoxy)phenyl]ethanone
SMIL CC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3

Tekniske data

Kemisk navn eller materiale 3',5'-Dibenzyloxyacetophenone
Smeltepunkt 60°C to 62°C
Mængde 10 g
Synonym 3',5'-dibenzyloxyacetophenone, 3,5-dibenzyloxyacetophenone, ethanone, 1-3,5-bis phenylmethoxy phenyl, 3,5-bis benzyloxy acetophenone, 1-3,5-bis phenylmethoxy phenyl ethanone, 1-3,5-bis benzyloxy phenyl ethanone, 3',5'-bis benzyloxy acetophenone, ethanone,1-3,5-bis phenylmethoxy phenyl
Opløselighedsinformation Soluble in dichloromethane,ethyl acetate and methanol
Formel vægt 332.4
Procent renhed 98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.