missing translation for 'onlineSavingsMsg'
Få mere at vide

3,4-dibrombenzaldehyd, 99 %, Thermo Scientific Chemicals

Product Code. 11353227
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
1 g
5 g
missing translation for 'unitSize'
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
11353227 5 g 5g
11343227 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11353227 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L13762.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74003-55-7 | C7H4Br2O | 263.916 g/mol

3,4-Dibrombenzaldehyd anvendes til fremstilling af isomert forgrenet pi-konjugeret system med terminal alkyn ved at reagere med 4-(triisopropylsilylethynyl) phenylacetylen.

Denne Thermo Scientific Chemicals mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific Chemicals .

Anvendelser 3,4-dibrombenzaldehyd anvendes til fremstilling af
isomert forgrenet pi-konjugeret system med terminal alkyn ved at reagere med 4-(triisopropylsilylethynyl) phenylacetylen.

Opløselighed Uopløseligt i
vand.

Bemærkninger
Luftfølsom. Uforeneligt med stærke oxidationsmidler.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74003-55-7
Molekylær formel C7H4Br2O
Molekylvægt (g/mol) 263.916
MDL nummer MFCD00236033
InChI nøgle UTYRZXNEFUYFCJ-UHFFFAOYSA-N
PubChem CID 622053
IUPAC navn 3,4-dibrombenzaldehyd
SMIL C1=CC(=C(C=C1C=O)Br)Br

Specifications

Kemisk navn eller materiale 3,4-Dibromobenzaldehyde
Smeltepunkt 69°C to 72°C
Mængde 5 g
Beilstein 2573433
Følsomhed Air Sensitive
Synonym 3,4-dibromo-benzaldehyde, benzaldehyde, 3,4-dibromo, benzaldehyde,3,4-dibromo, 1,2-dibromo-4-formylbenzene
Opløselighedsinformation Insoluble in water.
Formel vægt 263.93
Procent renhed 99%

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.