Få mere at vide
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-en-1-one, TRC
Artikelnummer. 30424765
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424765 100 mg
30409933 50 mg
30411862 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424765 Leverandør Toronto Research Chemicals Leverandørnr. TRCT303070100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1370008-65-3
Molekylær formel C11H17BO3
Molekylvægt (g/mol) 208.06
Synonym 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-cyclopenten-1-one, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-enone
IUPAC navn 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-en-1-one
SMIL CC1(C)OB(OC1(C)C)C2=CC(=O)CC2
Tekniske data
Kemisk navn eller materiale 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopent-2-en-1-one
Mængde 100 mg
InChI formel InChI=1S/C11H17BO3/c1-10(2)11(3,4)15-12(14-10)8-5-6-9(13)7-8/h7H,5-6H2,1-4H3
Formel vægt 208.1271