Få mere at vide
3-[4-(2-Methoxyphenyl)piperazin-1-yl]propan-1-amine, TRC
Artikelnummer. 30138009
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138009 50 mg
30136399 10 mg
30010185 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138009 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22939050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20529-23-1
Molekylær formel C14H23N3O
Molekylvægt (g/mol) 249.35
Synonym 1-Piperazinepropanamine, 4-(2-methoxyphenyl)-, Piperazine, 1-(3-aminopropyl)-4-(o-methoxyphenyl)- (8CI), 4-(2-Methoxyphenyl)-1-piperazinepropanamine, 1-(3-Aminopropyl)-4-(2-methoxyphenyl)piperazine, 1-(3-Aminopropyl)-4-(o-methoxyphenyl)piperazine, 1-Amino-3-[4-(2-methoxyphenyl)piperazin-1-yl]propane, 3-[4-(2-Methoxyphenyl)-1-piperazinyl]propylamine, 3-[4-(2-Methoxyphenyl)piperazino)propylamine, 4-(3-Aminopropyl)-1-(2-methoxyphenyl)piperazine
IUPAC navn 3-[4-(2-methoxyphenyl)piperazin-1-yl]propan-1-amine
SMIL COc1ccccc1N2CCN(CCCN)CC2
Tekniske data
Kemisk navn eller materiale 3-[4-(2-Methoxyphenyl)piperazin-1-yl]propan-1-amine
Mængde 50 mg
InChI formel InChI=1S/C14H23N3O/c1-18-14-6-3-2-5-13(14)17-11-9-16(10-12-17)8-4-7-15/h2-3,5-6H,4,7-12,15H2,1H3
Formel vægt 249.1841