Få mere at vide
3-(3-Aminobenzyl)phenylamine, TRC
Artikelnummer. 30007028
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007028 100 mg
30004958 10 mg
30005870 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007028 Leverandør Toronto Research Chemicals Leverandørnr. TRCA100820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19471-12-6
Molekylær formel C13H14N2
Molekylvægt (g/mol) 198.27
IUPAC navn 3-[(3-aminophenyl)methyl]aniline
SMIL Nc1cccc(Cc2cccc(N)c2)c1
Tekniske data
Kemisk navn eller materiale 3-(3-Aminobenzyl)phenylamine
Mængde 100 mg
InChI formel InChI=1S/C13H14N2/c14-12-5-1-3-10(8-12)7-11-4-2-6-13(15)9-11/h1-6,8-9H,7,14-15H2
Formel vægt 198.1157