Learn More
Sartorius™ 3.2 Standard Flexboy™ with TPE tubes
Designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products.
Brand: Sartorius™ FFB110912
Additional Details : Weight : 5.00000kg
Description
- Single-use, disposable
- Supplied sterilised and ready to use
Specifications
20 L | |
preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products | |
3 Ports |
Single-use | |
S71 | |
Gamma Irradiated |